Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(+)-2,2-Dimethyl-1,3-dioxolan-4-ylmethyl p-Toluenesulfonate 98.0+%, TCI America™
SDP

Catalog No. D25501G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D25501G 1 g
D25505G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D25501G Supplier TCI America Supplier No. D25501G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 23735-43-5
Molecular Formula C13H18O5S
Molecular Weight (g/mol) 286.342
MDL Number MFCD00063234
InChI Key SRKDUHUULIWXFT-NSHDSACASA-N
Synonym s-+-2,2-dimethyl-1,3-dioxolan-4-ylmethyl p-toluenesulfonate, s-2,2-dimethyl-1,3-dioxolan-4-yl methyl 4-methylbenzenesulfonate, 4s-2,2-dimethyl-1,3-dioxolan-4-yl methyl 4-methylbenzenesulfonate, s-+-2,2-dimethyl-1,3-dioxolane-4-ylmethyl p-toluenesulfonate, s-2,2-dimethyl-1,3-dioxolane-4-methanol p-toluenesulfonate, r---2,2-dimethyl-1,3-dioxolane-4-ylmethyl-p-toluenesulfonate, pubchem6795, 2,2-dimethyl-1,3-dioxolan-4-yl methyl 4-methylbenzenesulfonate #, s-2,2-dimethyl-1,3-dioxolan-4-ylmethyl p-toluenesulfonate
PubChem CID 2723760
IUPAC Name [(4S)-2,2-dimethyl-1,3-dioxolan-4-yl]methyl 4-methylbenzenesulfonate
SMILES CC1=CC=C(C=C1)S(=O)(=O)OCC2COC(O2)(C)C

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 286.34
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material (S)-(+)-2,2-Dimethyl-1,3-dioxolan-4-ylmethyl p-Toluenesulfonate
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.