Learn More
Cayman Chemical S 2 4DInItrophenylGlutath 10mg
SDP
Supplier:  Cayman Chemical 2466010 MG
Encompass_Preferred
A substrate for glutathione-S-transferase with a kinetic dissociation constant of 7 µM; transported by MDR1 and MDR3
Catalog No. 50-293-4868