Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(-)-2-Acetoxypropionyl Chloride 97.0+%, TCI America™
SDP

Catalog No. A12971G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A12971G 1 g
A12975G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A12971G Supplier TCI America Supplier No. A12971G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 36394-75-9
Molecular Formula C5H7ClO3
Molecular Weight (g/mol) 150.56
MDL Number MFCD00145252
InChI Key ALHZEIINTQJLOT-UHFFFAOYNA-N
Synonym (S)-(-)-O-Acetyllactoyl Chloride
PubChem CID 11094804
IUPAC Name 1-chloro-1-oxopropan-2-yl acetate
SMILES CC(OC(C)=O)C(Cl)=O

Specifications

Color Yellow
Boiling Point 78°C
Quantity 1 g
UN Number 3265
Formula Weight 150.56
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material (S)-(-)-2-Acetoxypropionyl Chloride
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.