Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(-)-2-Bromopropionic Acid 98.0+%, TCI America™
SDP

Catalog No. B175725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B175725G 25 g
B17575G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B175725G Supplier TCI America Supplier No. B175725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 32644-15-8
Molecular Formula C3H5BrO2
Molecular Weight (g/mol) 152.975
MDL Number MFCD00137325
InChI Key MONMFXREYOKQTI-REOHCLBHSA-N
Synonym s---2-bromopropionic acid, s-2-bromopropionic acid, 2s-2-bromopropanoic acid, s-2-bromopropanoic acid, l-2-bromopropionic acid, s---2-bromopropionicacid, l-alpha-bromopropionic acid, 2s---2-bromopropionic acid, s-alpha-bromopropionic acid
PubChem CID 642232
IUPAC Name (2S)-2-bromopropanoic acid
SMILES CC(C(=O)O)Br

Specifications

Melting Point -35°C
Boiling Point 94°C
Quantity 25 g
UN Number 3265
Formula Weight 152.98
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material (S)-(-)-2-Bromopropionic Acid
EINECSNumber (9)-1167
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.