Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(+)-2-Hydroxy-2-phenylpropionic acid, 98+%

Catalog No. AAL0869203
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0869203 1 g
AAL08692MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0869203 Supplier Thermo Scientific Chemicals Supplier No. L0869203
Add to Cart

CAS: 13113-71-8 | C9H10O3 | 166.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13113-71-8
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.18
MDL Number MFCD00067700
InChI Key NWCHELUCVWSRRS-UHFFFAOYNA-N
Synonym s-atrolactic acid, 2s-2-hydroxy-2-phenylpropanoic acid, s-+-2-hydroxy-2-phenylpropionic acid, atrolactic acid, +, s-2-hydroxy-2-phenylpropanoic acid, unii-22y6g519ru, s-+-atrolactic acid, atrolactic acid 2-phenyl-lactic acid, benzeneacetic acid, a-hydroxy-a-methyl-, as
PubChem CID 445144
ChEBI CHEBI:40741
IUPAC Name (2S)-2-hydroxy-2-phenylpropanoic acid
SMILES CC(O)(C(O)=O)C1=CC=CC=C1

Specifications

Melting Point 115°C to 117°C
Quantity 1 g
Beilstein 2208529
Optical Rotation +49° (c=2 in Water)
Formula Weight 166.18
Percent Purity ≥98%
Chemical Name or Material (S)-(+)-2-Hydroxy-2-phenylpropionic acid
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.