Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-2-Hydroxy-3-methoxy-3,3-diphenylpropionic Acid 98.0+%, TCI America™
SDP

Catalog No. H15681G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H15681G 1 g
H15685G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H15681G Supplier TCI America Supplier No. H15681G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 178306-52-0
Molecular Formula C16H16O4
Molecular Weight (g/mol) 272.3
MDL Number MFCD11113284
InChI Key RQJWOLFMWKZKCJ-CQSZACIVSA-N
PubChem CID 15380898
IUPAC Name (2S)-2-hydroxy-3-methoxy-3,3-diphenylpropanoic acid
SMILES COC(C1=CC=CC=C1)(C2=CC=CC=C2)C(C(=O)O)O

Specifications

Melting Point 120°C
Color White
Quantity 1 g
Formula Weight 272.30
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material (S)-2-Hydroxy-3-methoxy-3,3-diphenylpropionic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.