Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(+)-2-Methoxy-2-(1-naphthyl)propionic Acid 99.0+%, TCI America™
SDP

Catalog No. M1367100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1367100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1367100MG Supplier TCI America Supplier No. M1367100MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 102691-93-0
Molecular Formula C14H14O3
Molecular Weight (g/mol) 230.263
MDL Number MFCD03093634
InChI Key YVWMPILNFZOQSZ-AWEZNQCLSA-N
Synonym (S)-(+)-alpha-Methoxy-alpha-methyl-1-naphthaleneacetic Acid, (S)-(+)-M alpha NP Acid
PubChem CID 9834528
IUPAC Name (2S)-2-methoxy-2-naphthalen-1-ylpropanoic acid
SMILES CC(C1=CC=CC2=CC=CC=C21)(C(=O)O)OC

Specifications

Melting Point 110°C
Color White-Yellow
Quantity 100 mg
Formula Weight 230.26
Percent Purity ≥99.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material (S)-(+)-2-Methoxy-2-(1-naphthyl)propionic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.