Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(+)-2-Methoxypropanol, 97%, Acros Organics

Click to view available options
Quantity:
1mL
2.5mL
10mL
Packaging:
Glass bottle
116422-39-0
C4H10O2
90.12
MFCD01632593
YTTFFPATQICAQN-BYPYZUCNSA-N
2s-2-methoxypropan-1-ol,s-+-2-methoxypropanol,s-2-methoxypropan-1-ol,unii-6725gr28fg,2-methoxy-1-propanol, 2s,1-propanol, 2-methoxy-, 2s,2s-2-methoxy-1-propanol,s-2-methoxy-1-propanol,2-s-methoxypropan-1-ol,1-propanol, 2-methoxy-, s
642402
(2S)-2-methoxypropan-1-ol
CC(CO)OC
116422-39-0
C4H10O2
90.12
MFCD01632593
YTTFFPATQICAQN-BYPYZUCNSA-N
2s-2-methoxypropan-1-ol,s-+-2-methoxypropanol,s-2-methoxypropan-1-ol,unii-6725gr28fg,2-methoxy-1-propanol, 2s,1-propanol, 2-methoxy-, 2s,2s-2-methoxy-1-propanol,s-2-methoxy-1-propanol,2-s-methoxypropan-1-ol,1-propanol, 2-methoxy-, s
642402
(2S)-2-methoxypropan-1-ol
CC(CO)OC
Quantity 10mL
Water 0.1% max. (K.F.)
Density 0.9380g/mL
Specific Rotation Condition + 39.20 (20.00°C c=1.66,CHCl3)
Refractive Index 1.4060 to 1.4080
Specific Rotation + 39.20
Specific Gravity 0.938
Packaging Glass bottle
Flash Point 33°C
Color Colorless
Infrared Spectrum Authentic
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Linear Formula CH3CH(OCH3)CH2OH
Formula Weight 90.12
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material (S)-(+)-2-Methoxypropanol, 99%
Show More Show Less

GHS H Statement
Flammable liquid and vapor.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.

GHS Signal Word: Warning

TSCA : TSCA