Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-2-Methylbutyl p-Toluenesulfonate 95.0+%, TCI America™
SDP

Catalog No. T159525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T159525G 25 g
T15955G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T159525G Supplier TCI America Supplier No. T159525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 38261-81-3
Molecular Formula C12H18O3S
Molecular Weight (g/mol) 242.33
MDL Number MFCD00013399
InChI Key HPEVJTNZYIMANV-UHFFFAOYNA-N
Synonym p-Toluenesulfonic Acid (S)-2-Methylbutyl Ester
PubChem CID 113324
IUPAC Name 2-methylbutyl 4-methylbenzene-1-sulfonate
SMILES CCC(C)COS(=O)(=O)C1=CC=C(C)C=C1

Specifications

Color Yellow
Quantity 25 g
Formula Weight 242.33
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material (S)-2-Methylbutyl p-Toluenesulfonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.