Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(+)-2-Methylpiperazine 97.0+%, TCI America™
SDP

Catalog No. M14241G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M14241G 1 g
M14245G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M14241G Supplier TCI America Supplier No. M14241G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 74879-18-8
Molecular Formula C5H12N2
Molecular Weight (g/mol) 100.165
MDL Number MFCD00171452
InChI Key JOMNTHCQHJPVAZ-YFKPBYRVSA-N
Synonym s-+-2-methylpiperazine, s-2-methylpiperazine, 2s-2-methylpiperazine, piperazine, 2-methyl-, 2s, s-2-methyl-piperazine, unii-c5vi3s1yc0, 2-methylpiperazine, s, 2-s-methylpiperazine, s-mp, c5vi3s1yc0
PubChem CID 2734219
IUPAC Name (2S)-2-methylpiperazine
SMILES CC1CNCCN1

Specifications

Melting Point 91°C
Color White-Yellow
Quantity 1 g
UN Number 2925
Formula Weight 100.17
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material (S)-(+)-2-Methylpiperazine
EINECSNumber (5)-3829
RTECSNumber TM1225020
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.