Learn More
Beckman Coulter Swinging Bucket Rotor, SX4750, for Allegra X-15/X-15R Centrifuges
Supplier:  Beckman Coulter 361111
Encompass
The Beckman Coulter SW 32 Ti Swinging-Bucket Rotor is designed for high-capacity centrifugation needs. This rotor holds 6 x 750mL bottles and can reach speeds up to 22,000 rpm with a maximum RCF of 48,384 x g. The titanium construction provides corrosion resistance while maintaining high strength. The rotor has a load capacity of 30kg and is compatible with multiple Beckman Coulter floor model ultracentrifuges including the Optima XE/XP and Avanti J-E centrifuges. The advanced design reduces vibration and increases sample recovery.
Catalog No. NC1010380