Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(-)-3,3'-Di-tert-butyl-5,5',6,6'-tetramethylbiphenyl-2,2'-diol, 99%

Catalog No. AAH2734203
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2734203 1 g
AAH27342MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2734203 Supplier Thermo Scientific Chemicals Supplier No. H2734203
Add to Cart

CAS: 205927-03-3 | C24H34O2 | 354.53 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 205927-03-3
Molecular Formula C24H34O2
Molecular Weight (g/mol) 354.53
MDL Number MFCD01862439
InChI Key NMVVBVMYPLMIOU-UHFFFAOYSA-N
Synonym 3,3'-di-tert-butyl-5,5',6,6'-tetramethyl-1,1'-biphenyl-2,2'-diol, 5,5',6,6'-tetramethyl-3,3'-di-tert-butyl-1,1'-biphenyl-2,2'-diol, 3,3'-di-t-butyl-5,5',6,6'-tetramethyl-1,1'-biphenyl-2,2'-diol, s---3,3'-di-tert-butyl-5,5',6,6'-tetramethylbiphenyl-2,2'-diol, 6-tert-butyl-2-3-tert-butyl-2-hydroxy-5,6-dimethylphenyl-3,4-dimethylphenol, 2,2'-bi 6-tert-butyl-3,4-dimethylphenol, r-3,3'-di-tert-butyl-5,5',6,6'-tetramethylbiphenyl-2,2'-diol, s-3,3'-di-tert-butyl-5,5',6,6'-tetramethylbiphenyl-2,2'-diol, as-3,3'-di-tert-butyl-5,5',6,6'-tetramethylbiphenyl-2,2'-diol, s--5,5,6,6-tetramethyl-3,3-di-tert-butyl-1,1-biphenyl-2,2-diol
PubChem CID 3255344
IUPAC Name 6-tert-butyl-2-(3-tert-butyl-2-hydroxy-5,6-dimethylphenyl)-3,4-dimethylphenol
SMILES CC1=CC(=C(O)C(=C1C)C1=C(C)C(C)=CC(=C1O)C(C)(C)C)C(C)(C)C

Specifications

Melting Point 171°C to 173°C
Quantity 1 g
Optical Rotation −84° (c=0.35 in THF)
Formula Weight 354.54
Percent Purity 99%
Chemical Name or Material (S)-(-)-3,3'-Di-tert-butyl-5,5',6,6'-tetramethylbiphenyl-2,2'-diol
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.