Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abcam (S)-3,4-DCPG, mGlu8a Agonist, 99% Purity, 50 mM/mL
Description
- Potency: Potent, selective mGlu8a agonist with an EC50 of 31 nM.
- Selectivity: Displays >100-fold selectivity over mGlu1-7.
- Chemical Properties: Molecular weight of 239.18 Da, purity >99%.
- Activity: Systemically active in vivo.
- Solubility: Soluble in 1 mL water to give the specified concentration.
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | mGlu8a |
| Shipping Condition | Ambient |
| Substrate Properties | Chemical Substrate |
| Format | Vial |
| Alternate Part Number | UBP1109 |
| Buffer | 100% (S)-3,4-Dicarboxyphenylglycine |
| Concentration | 50 mM |
| Content And Storage | Store at 4°C. |
| Form | Solution |
| Formulation | 100% (S)-3,4-Dicarboxyphenylglycine |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?