Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-3-Aminotetrahydrofuran hydrochloride, 95%, Thermo Scientific™

Catalog No. AC456930010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC456930010 1 g
AC456930050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC456930010 Supplier Thermo Scientific Chemicals Supplier No. 456930010

Chemical Identifiers

CAS 204512-95-8
Molecular Formula C4H9NO·ClH
Molecular Weight (g/mol) 123.58
InChI Key MHOVLDXJDIEEMJ-WCCKRBBISA-N
Synonym s-tetrahydrofuran-3-amine hydrochloride, s-3-aminotetrahydrofuran hydrochloride, 3s-oxolan-3-amine hydrochloride, s-3-aminotetrahydrofuran hcl, s-tetrahydrofuran-3-amine hcl, s-tetrahydro-furan-3-ylamine hydrochloride, 3-furanamine, tetrahydro-, hydrochloride, 3s, s-3-aminotetrahydrofuranhydrochloride, pubchem15126, ksc916a5t
PubChem CID 18664284
IUPAC Name (3S)-oxolan-3-amine;hydrochloride
SMILES C1COCC1N.Cl

Specifications

Melting Point 140.0°C to 145.0°C
Infrared Spectrum Authentic
Assay Percent Range 94.0 to max. 106% min. (Argentometry)
Packaging Glass Bottle
Quantity 1 g
Specific Rotation -7.4
Formula Weight 123.58
Percent Purity 95%
Chemical Name or Material (S)-3-Aminotetrahydrofuran hydrochloride
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
Precautionary Statement GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Call a POISON CENTER or doctor/physician if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.