Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-3-(Benzoylthio)-2-methylpropionic Acid 98.0+%, TCI America™
SDP

Catalog No. B53155G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B53155G 5 g
B53151G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B53155G Supplier TCI America Supplier No. B53155G

Chemical Identifiers

CAS 72679-02-8
Molecular Formula C11H12O3S
Molecular Weight (g/mol) 224.274
MDL Number MFCD00038376
InChI Key BCAYPPFBOJCRPN-MRVPVSSYSA-N
PubChem CID 12763916
IUPAC Name (2S)-3-benzoylsulfanyl-2-methylpropanoic acid
SMILES CC(CSC(=O)C1=CC=CC=C1)C(=O)O

Specifications

Melting Point 69°C
Color White-Yellow
Quantity 5 g
Formula Weight 224.27
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material (S)-3-(Benzoylthio)-2-methylpropionic Acid
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.