Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-3-(Dimethylamino)-1-(2-thienyl)-1-propanol 98.0+%, TCI America™
SDP

Catalog No. D40101G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D40101G 1 g
D40105G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D40101G Supplier TCI America Supplier No. D40101G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 132335-44-5
Molecular Formula C9H15NOS
Molecular Weight (g/mol) 185.29
MDL Number MFCD07782107
InChI Key XWCNSHMHUZCRLN-UHFFFAOYNA-N
Synonym (S)-2-[3-(Dimethylamino)-1-hydroxypropyl]thiophene
PubChem CID 9964277
IUPAC Name 3-(dimethylamino)-1-(thiophen-2-yl)propan-1-ol
SMILES CN(C)CCC(O)C1=CC=CS1

Specifications

Melting Point 79°C
Color White
Quantity 1 g
Formula Weight 185.29
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material (S)-3-(Dimethylamino)-1-(2-thienyl)-1-propanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.