Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-3-Phenylcyclohexanone 98.0+%, TCI America™
SDP

Catalog No. P1500100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P1500100MG 100 mg
P15001G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P1500100MG Supplier TCI America Supplier No. P1500100MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 57344-86-2
Molecular Formula C12H14O
Molecular Weight (g/mol) 174.24
MDL Number MFCD08276411
InChI Key CJAUDSQXFVZPTO-UHFFFAOYNA-N
PubChem CID 11116555
IUPAC Name 3-phenylcyclohexan-1-one
SMILES O=C1CCCC(C1)C1=CC=CC=C1

Specifications

Color White
Quantity 100 mg
Formula Weight 174.24
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material (S)-3-Phenylcyclohexanone
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.