Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(-)-4-Benzyl-2-oxazolidinone, 98+%, Thermo Scientific™

Catalog No. AA4181806
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4181806 5 g
AA4181803 1 g
AA4181814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4181806 Supplier Thermo Scientific Chemicals Supplier No. 4181806

Used as chiral auxiliary during asymmetric synthesis

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90719-32-7
Molecular Formula C10H11NO2
Molecular Weight (g/mol) 177.20
MDL Number MFCD00064496
InChI Key OJOFMLDBXPDXLQ-VIFPVBQESA-N
Synonym s-4-benzyl-2-oxazolidinone, s-4-benzyloxazolidin-2-one, 4s-4-benzyl-1,3-oxazolidin-2-one, s---4-benzyl-2-oxazolidinone, s-4-benzyl-oxazolidin-2-one, 2-oxazolidinone, 4-phenylmethyl-, 4s, s-+-4-benzyl-2-oxazolidinone, s-4-phenylmethyl-2-oxazolidinone, 4s-4-phenylmethyl-1,3-oxazolidin-2-one
PubChem CID 736225
IUPAC Name (4S)-4-benzyl-1,3-oxazolidin-2-one
SMILES O=C1N[C@@H](CC2=CC=CC=C2)CO1

Specifications

Melting Point 89°C to 90°C
Quantity 5 g
Formula Weight 177.2
Percent Purity ≥98%
Physical Form Powder
Chemical Name or Material (S)-(-)-4-Benzyl-2-oxazolidinone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.