Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-4-Benzyloxy-1,2-butanediol 95.0+%, TCI America™
SDP

Catalog No. B29001G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B29001G 1 g
B2900200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B29001G Supplier TCI America Supplier No. B29001G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 69985-32-6
Molecular Formula C11H16O3
Molecular Weight (g/mol) 196.25
MDL Number MFCD10000950
InChI Key TVRPDIKPMQUOSL-UHFFFAOYNA-N
PubChem CID 9794180
IUPAC Name 4-(benzyloxy)butane-1,2-diol
SMILES OCC(O)CCOCC1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 170°C
Quantity 1 g
Formula Weight 196.25
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material (S)-4-Benzyloxy-1,2-butanediol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.