Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-6,6'-Dibromo-2,2'-bis(methoxymethoxy)-1,1'-binaphthyl 98.0+%, TCI America™
SDP

Catalog No. D48835G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D48835G 5 g
D48831G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D48835G Supplier TCI America Supplier No. D48835G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 211560-97-3
Molecular Formula C24H20Br2O4
Molecular Weight (g/mol) 532.228
MDL Number MFCD03788936
InChI Key UPLLZDVWXACTEG-UHFFFAOYSA-N
PubChem CID 10839989
IUPAC Name 6-bromo-1-[6-bromo-2-(methoxymethoxy)naphthalen-1-yl]-2-(methoxymethoxy)naphthalene
SMILES COCOC1=C(C2=C(C=C1)C=C(C=C2)Br)C3=C(C=CC4=C3C=CC(=C4)Br)OCOC

Specifications

Melting Point 134°C
Color White-Yellow
Quantity 5 g
Formula Weight 532.23
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material (S)-6,6′-Dibromo-2,2′-bis(methoxymethoxy)-1,1′-binaphthyl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.