Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(+)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetyl chloride, 98+%

Catalog No. AAL1433203
Change view
Click to view available options
Quantity:
50 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1433203 1 g
AAL14332MF 50 mg
AAL14332MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1433203 Supplier Thermo Scientific Chemicals Supplier No. L1433203
Add to Cart

CAS: 20445-33-4 | C10H8ClF3O2 | 252.617 g/mol

(S)-(+)-α-Methoxy-α-(trifluoromethyl)phenylacetyl chloride is used in the preparation of natural products and pheromones. It is also employed as a derivatization enantiomeric purity reagent used for the determination of enantiomeric purity of alcohol and amines by gas chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-(+)-α-Methoxy-α-(trifluoromethyl)phenylacetyl chloride is used in the preparation of natural products and pheromones. It is also employed as a derivatization enantiomeric purity reagent used for the determination of enantiomeric purity of alcohol and amines by gas chromatography.

Solubility
Slightly miscible with water.

Notes
Moisture sensitive. Store in a cool place. Incompatible with strong oxidizing agents, strong acids and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20445-33-4
Molecular Formula C10H8ClF3O2
Molecular Weight (g/mol) 252.617
MDL Number MFCD00067105
InChI Key PAORVUMOXXAMPL-SECBINFHSA-N
Synonym s-3,3,3-trifluoro-2-methoxy-2-phenylpropanoyl chloride, +-mtpa-cl, unii-472va70ul4, 2s-3,3,3-trifluoro-2-methoxy-2-phenylpropanoyl chloride, s-+-alpha-methoxy-alpha-trifluoromethylphenylacetyl chloride, s-+-a-methoxy-a-trifluoromethylphenylacetyl chloride, s-+-alpha-methoxy-alpha-trifluoromethylphenylacetylchloride, s-+-alpha-methoxy-alpha-trifluoromethyl phenylacetyl chloride, +-mtpa chloride, +-mosher's chloride
PubChem CID 2724611
IUPAC Name (2S)-3,3,3-trifluoro-2-methoxy-2-phenylpropanoyl chloride
SMILES COC(C1=CC=CC=C1)(C(=O)Cl)C(F)(F)F

Specifications

Density 1.353
Boiling Point 213°C to 214°C
Flash Point 89°C (192°F)
Refractive Index 1.469
Quantity 1 g
UN Number UN3265
Beilstein 3591564
Sensitivity Moisture sensitive
Merck Index 14,6280
Solubility Information Slightly miscible with water.
Optical Rotation +137° (c=6.4 in Carbon Tetrachloride)
Formula Weight 252.62
Percent Purity ≥98%
Chemical Name or Material (S)-(+)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetyl chloride
Show More Show Less
Hazard Category H227-H314-H335
Hazard Statement GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement P210-P235-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Store at -20°C; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.