Learn More
AdipoGen S Ascochin
SDP
Supplier:  AdipoGen BVT0309M005
Encompass_Preferred
Chemical. CAS 935699-58-4. Formula C12H10O5. MW 234.2. Isolated from Ascochyta sp. Antibiotic. Antifungal and phytotoxic.
Catalog No. 50-256-1996
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.