Learn More
S-Curve™ Four Compartment Glove Dispenser with Front Access Holes
Catalog No. 19160200 Shop All S-Curve Technologies Products
Change view
Click to view available options
Material:
Acrylic
PETG
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
19-160-200 Acrylic
19-162-430 PETG
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-160-200 Supplier S-Curve™ Supplier No. GD410
Add to Cart
Add to Cart
This item is not returnable. View return policy
Includes
Heavy-duty wall bracket and support bar, mounting instructions.
Dispenser features front access holes for easy product dispensing and a single, sloping lid with full-length hinge to maintain HEPA flow.
  • Single, sloping lid with full-length hinge to maintain HEPA flow
  • Front access holes for easy product dispensing
  • Can be mounted with heavy-duty wall brackets (included)
  • Flame treated edges for a smooth finish
Specifications
Product Type Glove Dispenser
Color Clear
Dimensions (D x W x H) Interior 12 x 21 x 12 in.
Holds Gloves
Material Acrylic
Mounting Keyhole Slots
No. of Compartments 4
Product Style Top Loading
Includes Mounting hardware
Thickness (English) 0.25 in.