Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S-Curve™ EBD-XUP Series Exam Glove Box Dispenser

Catalog No. 19162404 Shop All S-Curve Technologies Products
Change view
Click to view available options
Capacity:
1 Box
2 Boxes
3 Boxes
4 Boxes
Dimensions (D x W x H) Interior:
4 x 5.75 x 10 in.
4 x 11.4 x 10 in.
4 x 22.6 x 10 in.
4 x 17 x 10 in.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity Dimensions (D x W x H) Interior
19-162-404 1 Box 4 x 5.75 x 10 in.
19-162-405 2 Boxes 4 x 11.4 x 10 in.
19-162-406 3 Boxes 4 x 17 x 10 in.
19-999-425 4 Boxes 4 x 22.6 x 10 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-162-404 Supplier S-Curve™ Supplier No. EBD1UP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Dispensers are constructed of 1/8 in. clear PETG and feature individual compartments to keep multiple glove boxes neat and organized.

  • Constructed of 1/8 in. clear PETG material
  • Keyholes on the back for wall mounting with included hardware
  • Dispensers can be mounted horizontally or vertically
  • Each individual compartment measures 3.75 x 5.5 x 10 in.
  • Flame polished edges for a smooth finish

Specifications

Product Type Glove Box Holder
Color Clear
Dimensions (D x W x H) Interior 4 x 5.75 x 10 in.
Holds Exam Glove Boxes
Material PETG
Mounting Keyhole Slots
No. of Compartments 1
Capacity 1 Box
Includes Mounting hardware
Thickness (English) 0.125 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.