Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S-Curve™ X-Large Bouffant Dispenser

Catalog No. 19162567 Shop All S-Curve Technologies Products
Change view
Click to view available options
Material:
PETG
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
19-162-567 PETG
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-162-567 Supplier S-Curve™ Supplier No. BD616PETG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Product Type Bouffant Cap Dispenser
Color Clear
Dimensions (D x W x H) Interior 6 x 12 x 16 in.
Material PETG
Mounting Free-standing or Wall Mount
No. of Compartments 1
Product Style Hinged Lid with Oval Front Opening
Edges Flame Treated
For Use With (Application) Holds large volumes of bouffants, beard covers, etc
Size X-Large
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.