Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S-Curve™ Three Chamber Finger Cot Dispenser

Catalog No. 19162579 Shop All S-Curve Technologies Products
Change view
Click to view available options
Material:
Acrylic
PETG
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
19-162-579 PETG
19-162-590 Acrylic
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-162-579 Supplier S-Curve™ Supplier No. FCD03PETG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Dispensers feature three compartments with access trays and lid. Flame-treated edges ensure a smooth finish, with rubber footing to prevent scratching.

  • Three compartments with access trays and lid
  • Flame-treated edges for a smooth finish
  • Rubber padding on bottom corners to prevent scratching/slipping
  • Can be mounted to the wall or free standing

Specifications

Product Type Finger Cot Dispenser
Color Clear
Dimensions (D x W x H) Interior 10 x 17 x 12 in.
Holds Finger Cots, Ear Plugs
Material PETG
Mounting Keyhole Slots
No. of Compartments 3
Product Style Top Loading
Thickness (English) 0.25 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.