Learn More
Chemscene ChemScene | (S)-Fmoc-2-amino-5-(tritylthio)-pentanoic acid | 1G | CS-0531924 | 0.98 | 1417789-17-3| MFCD21364742 | 613.76
SDP
Supplier:  Chemscene CS05319241G
Encompass_Preferred
ChemScene | (S)-Fmoc-2-amino-5-(tritylthio)-pentanoic acid | 1G | CS-0531924 | 0.98 | 1417789-17-3| MFCD21364742 | 613.76
Catalog No. NC2431850
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.