Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S-Furfuryl Thioacetate 97.0+%, TCI America™
SDP

Catalog No. T128310G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T128310G 10 g
T128325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T128310G Supplier TCI America Supplier No. T128310G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 13678-68-7
Molecular Formula C7H8O2S
Molecular Weight (g/mol) 156.199
MDL Number MFCD00010083
InChI Key LQOUTUIIYXYBQW-UHFFFAOYSA-N
Synonym s-furfuryl thioacetate, furfuryl thioacetate, furfurylthiol acetate, ethanethioic acid, s-2-furanylmethyl ester, s-furfuryl ethanethioate, acetic acid, thio-, s-furfuryl ester, s-2-furanylmethyl ethanethioate, fema no. 3162, 2-furfurylthiol acetate, thioacetic acid s-furfuryl ester
PubChem CID 61660
IUPAC Name S-(furan-2-ylmethyl) ethanethioate
SMILES CC(=O)SCC1=CC=CO1

Specifications

Boiling Point 91°C
Quantity 10 g
Formula Weight 156.20
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material S-Furfuryl Thioacetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.