Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-Glycidyl Butyrate 98.0+%, TCI America™
SDP

Catalog No. G028325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G028325G 25 g
G02835G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G028325G Supplier TCI America Supplier No. G028325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 65031-96-1
Molecular Formula C7H12O3
Molecular Weight (g/mol) 144.17
MDL Number MFCD00273369
InChI Key YLNSNVGRSIOCEU-LURJTMIESA-N
Synonym s-+-glycidyl butyrate, s-glycidyl butyrate, s-oxiran-2-ylmethyl butyrate, 2s-oxiran-2-yl methyl butanoate, butanoic acid, 2s-oxiranylmethyl ester, s-glycidyl butanoate, pubchem5837, s-2-butyryloxymethyl oxirane, butyric acid s-glycidyl ester
PubChem CID 6999819
IUPAC Name [(2S)-oxiran-2-yl]methyl butanoate
SMILES CCCC(=O)OCC1CO1

Specifications

Color Yellow
Boiling Point 90°C
Quantity 25 g
Formula Weight 144.17
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material (S)-Glycidyl Butyrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.