Learn More
Enzo Life Sciences (S)-Ketoprofen (1g). CAS: 22161-81-5
SDP
Supplier:  Enzo Life Sciences BMLEI2880001
Encompass_Preferred
Active enantiomer of ketoprofen. Inhibits cyclooxygenase. Alternative name: (S)-(+)-3-Benzoyl-methylbenzene acetic acid. Purity: ≥98.5% (HPLC). Solubility: Soluble in 100% ethanol (25mg/ml). Long Term Storage: Ambient.
Catalog No. 50-201-0670
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.