Learn More
Medchemexpress LLC (S)-mandelic acid | 17199-29-0 | MFCD00004495 | 152.15 | C8H8O3 | 100g
SDP
Supplier:  Medchemexpress LLC HYY0816100G
Encompass_Preferred
(S)-Mandelic Acid is a (S)-enantiomer of Mandelic Acid (HY-W015591) (S)-Mandelic Acid undergoes one-directional chiral inversion to (R)-Mandelic Acid in rat models (S)-Mandelic acid is a biochemical reagent that can be used as a biological material or organic compound for life science related research[1]
Catalog No. 50-002-69826