Learn More
STEMCELL Technologies (S)-MG132, Size: 1 mg
SDP
Supplier:  STEMCELL Technologies 73262
Encompass
(S)-MG132 is a reversible, cell-permeable inhibitor of proteasome activity (IC₅₀ = 100 nM; Kisselev & Goldberg) and calpain (IC₅₀ = 1.2 µM; Tsubuki et al.). The ubiquitin-proteasome pathway selectively degrades intracellular proteins, thereby clearing damaged or misfolded proteins, and regulating the availability of key proteins involved in the control of inflammatory processes and cell cycle regulation. (S)-MG132 suppresses NF-κB activation by preventing IκB degradation (IC₅₀ = 3 µM; Arlt et al., Palombella et al., Ortiz-Lazareno et al.).CANCER RESEARCH· Blocks apoptosis triggered by DNA damage in HeLa cells (Zhang et al.).· Inhibits NF-κB activation, sensitizing a variety of carcinoma cell lines to apoptosis (Arlt et al.).· Cytotoxic effects on a variety of human cancer cell lines (Banerjee & Liefshitz).· Inhibits growth of mouse melanoma (B16) and human ocular melanoma (IPC227F) cell lines (Vivier et al.).
Catalog No. 50-197-6049
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.