Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-N-Boc-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, 98%

Catalog No. AAH6142303
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6142303 1 g
AAH6142306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6142303 Supplier Thermo Scientific Chemicals Supplier No. H6142303

CAS: 78879-20-6 | C15H18NO4 | 276.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78879-20-6
Molecular Formula C15H18NO4
Molecular Weight (g/mol) 276.31
MDL Number MFCD00143845
InChI Key HFPVZPNLMJDJFB-LBPRGKRZSA-M
Synonym boc-tic-oh, s-2-tert-butoxycarbonyl-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, s-n-boc-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, boc-l-tic-oh, s-2-boc-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, 3s-2-tert-butoxycarbonyl-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, n-boc-l-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, 3s-2-tert-butoxycarbonyl-3,4-dihydro-1h-isoquinoline-3-carboxylic acid, 3s-2-2-methylpropan-2-yl oxycarbonyl-3,4-dihydro-1h-isoquinoline-3-carboxylic acid
PubChem CID 664088
IUPAC Name (3S)-2-[(2-methylpropan-2-yl)oxycarbonyl]-3,4-dihydro-1H-isoquinoline-3-carboxylic acid
SMILES CC(C)(C)OC(=O)N1CC2=CC=CC=C2C[C@H]1C([O-])=O

Specifications

Color White
Quantity 1 g
Beilstein 4297114
Formula Weight 277.32
Percent Purity 98%
Physical Form Solid
Chemical Name or Material (S)-N-Boc-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.