Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-N-Glycidylphthalimide 98.0+%, TCI America™
SDP

Catalog No. G032825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G032825G 25 g
G03285G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G032825G Supplier TCI America Supplier No. G032825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 161596-47-0
Molecular Formula C11H9NO3
Molecular Weight (g/mol) 203.197
MDL Number MFCD04973350
InChI Key DUILGEYLVHGSEE-ZETCQYMHSA-N
Synonym (S)-N-(2,3-Epoxypropyl)phthalimide
PubChem CID 719412
IUPAC Name 2-[[(2S)-oxiran-2-yl]methyl]isoindole-1,3-dione
SMILES C1C(O1)CN2C(=O)C3=CC=CC=C3C2=O

Specifications

Melting Point 102°C
Color White
Quantity 25 g
Formula Weight 203.20
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (S)-N-Glycidylphthalimide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.