Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(-)-N,N-Dimethyl-1-phenylethylamine 98.0+%, TCI America™
SDP

Catalog No. D16871G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D16871G 1 g
D16875G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D16871G Supplier TCI America Supplier No. D16871G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 17279-31-1
Molecular Formula C10H15N
Molecular Weight (g/mol) 149.237
MDL Number MFCD00056236
InChI Key BVURNMLGDQYNAF-VIFPVBQESA-N
Synonym (S)-(-)-alpha-Methyl-N,N-dimethylbenzylamine
PubChem CID 2724276
IUPAC Name (1S)-N,N-dimethyl-1-phenylethanamine
SMILES CC(C1=CC=CC=C1)N(C)C

Specifications

Color Yellow
Boiling Point 71°C
Quantity 1 g
Formula Weight 149.24
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material (S)-(-)-N,N-Dimethyl-1-phenylethylamine
EINECSNumber (3)-0318
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.