Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(-)-Nicotine, 98+%

Catalog No. p-7082113
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1239814 25 g
AAA1239822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1239814 Supplier Thermo Scientific Chemicals Supplier No. A1239814
Only null left
Add to Cart

L-Nicotine, CAS # 54-11-5, has been shown in laboratory studies to alleviate smoking habits. It is also known as 3-(1-methyl-2-pyrrolidinyl)pyridine or (S)-(-)-nicotine. | CAS: 54-11-5 | C10H14N2 | 162.236 g/mol

(S)-(-)-Nicotine is utilized as a therapeutic agent to eliminate smoking with the damage it does to health. It is widely used as a drug to enhance the memory or other cognition functions, which is second to caffeine. Lesser levels of nicotine given in the form of gum, may help to eliminate smoking.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description
• L-Nicotine is a naturally occuring alkaloid in the nightshade family of plants
• It is also known as 3-(1-methyl-2-pyrrolidinyl)pyridine or (S)-(-)-nicotine
Applications
• L-Nicotine has been shown in studies to alleviate smoking habits
• It has also been shown to enhance cognitive functions such as memory
• Studies show that lower levels of nicotine in the form of gum may also help eliminate smoking habits

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54-11-5
Molecular Formula C10H14N2
MDL Number MFCD00006369
InChI Key SNICXCGAKADSCV-JTQLQIEISA-N
Synonym nicotine, l-nicotine, --nicotine, s-nicotine, habitrol, s-3-1-methylpyrrolidin-2-yl pyridine, nicoderm, nicotrol, s---nicotine, nicoderm cq
PubChem CID 89594
ChEBI CHEBI:17688
IUPAC Name 3-[(2S)-1-methylpyrrolidin-2-yl]pyridine

Specifications

Melting Point °C to 79°C
Density 1.017
Boiling Point 245°C to 248°C
Flash Point 101°C (213°F)
Assay Percent Range 98+%
Refractive Index 1.5265
Quantity 25 g
UN Number UN1654
Beilstein 82109
Sensitivity Air and light sensitive; Hygroscopic
Merck Index 14,6524
Solubility Information Miscible with water,alcohol,ether and light petroleum.
SMILES CN1CCCC1C2=CN=CC=C2
Molecular Weight (g/mol) 162.236
Formula Weight 162.24
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material (S)-(-)-Nicotine
Show More Show Less
Hazard Category H300+H310+H330
Hazard Statement GHS H Statement
H301-H310
Precautionary Statement P260-P262-P264b-P270-P271-P280-P284-P301+P310-P302+P350-P304+P340-P310-P312-P314-P330-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: NICOTINE
EINECSNumber 200-193-3
RTECSNumber QS5250000
TSCA Yes
storageNote1 Air Sensitive and Hygroscopic
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.