Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(+)-O-Acetylmandelic acid, 99%

Catalog No. AAH5656703
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5656703 1 g
AAH5656706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5656703 Supplier Thermo Scientific Chemicals Supplier No. H5656703
Add to Cart

CAS: 7322-88-5 | C10H10O4 | 194.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7322-88-5
Molecular Formula C10H10O4
Molecular Weight (g/mol) 194.19
MDL Number MFCD00064215,MFCD00662355,MFCD00004249
InChI Key OBCUSTCTKLTMBX-UHFFFAOYNA-N
Synonym s-2-acetoxy-2-phenylacetic acid, +-o-acetyl-l-mandelic acid, s-+-alpha-acetoxyphenylacetic acid, s-acetylmandelic acid, 2s-2-acetyloxy-2-phenylacetic acid, s-o-acetylmandelic acid, mandelic acid, acetate, l, unii-0n6kt18k98, l-+-o-acetylmandelic acid, s-+-o-acetyl-l-mandelic acid
PubChem CID 6923379
SMILES CC(=O)OC(C(O)=O)C1=CC=CC=C1

Specifications

Melting Point 97°C to 100°C
Quantity 1 g
Beilstein 2694109
Optical Rotation +153° (c=2 in Acetone)
IUPAC Name 2-(acetyloxy)-2-phenylacetic acid
Formula Weight 194.19
Percent Purity 99%
Chemical Name or Material (S)-(+)-O-Acetylmandelic acid
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.