Learn More
AdipoGen S-O-Desmethylnaproxen
SDP
Supplier:  AdipoGen CDXD0390M010
Encompass_Preferred
Chemical. CAS 52079-10-4. Formula C13H12O3. MW 216.2. Synthetic. Major naproxen demethylation metabolite. Naproxen demethylation is mediated primarily by CYP2C9. Can be used for assaying naproxen metabolites.
Catalog No. 50-256-1097
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.