Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(-)-Propylene Oxide 98.0+%, TCI America™
SDP

Catalog No. P095125ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 mL
5 mL
25 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P095125ML 25 mL
P09511ML 1 mL
P09515ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. P095125ML Supplier TCI America Supplier No. P095125ML

Chemical Identifiers

CAS 16088-62-3
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00064312
InChI Key GOOHAUXETOMSMM-VKHMYHEASA-N
Synonym s---propylene oxide, s-propylene oxide, 2s-2-methyloxirane, s-1,2-epoxypropane, --propylene oxide, s-2-methyloxirane, --methyloxirane, s-methyloxirane, s---1,2-epoxypropane
PubChem CID 146262
ChEBI CHEBI:28982
IUPAC Name (2S)-2-methyloxirane
SMILES C[C@H]1CO1

Specifications

Color Colorless
Boiling Point 34°C
Quantity 25 mL
UN Number 1280
Formula Weight 58.08
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material (S)-(-)-Propylene Oxide
EINECSNumber (2)-0219
RTECSNumber UJ2651000
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.