Learn More
Bon Opus Biosciences Recombinant S Protein RBD (Mammalian, D364Y, C-6His) -50ug
SDP
Supplier:  Bon Opus Biosciences DRA8150UG
Encompass_Preferred
Recombinant 2019-nCoV S protein RBD Protein is produced by our Mammalian expression system and the target gene encoding Arg319-Phe541(D364Y) is expressed with a 6His tag at the C-terminus. Expression system: Human Cells. Accession# QHD43416.1. Formulation: Supplied as a 0.2 μm filtered solution of PBS, pH 7.4. Bon Opus Cat# DRA81
Catalog No. 50-208-0571
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.