Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals (S,R)-Noscapine

Catalog No. AC462710050
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC462710050 5 g
AC462710250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC462710050 Supplier Thermo Scientific Chemicals Supplier No. 462710050
Add to Cart

CAS: 128-62-1 | C22H23NO7 | 413.43 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 128-62-1
Molecular Formula C22H23NO7
Molecular Weight (g/mol) 413.43
InChI Key AKNNEGZIBPJZJG-MSOLQXFVSA-N
IUPAC Name (3S)-6,7-dimethoxy-3-[(5R)-4-methoxy-6-methyl-2H,5H,6H,7H,8H-[1,3]dioxolo[4,5-g]isoquinolin-5-yl]-1,3-dihydro-2-benzofuran-1-one
SMILES COC1=CC=C2[C@H](OC(=O)C2=C1OC)[C@@H]1N(C)CCC2=CC3=C(OCO3)C(OC)=C12

Specifications

Melting Point 174.0°C to 177.0°C
Quantity 5 g
Solubility Information Solubility aq. soln.: practically insoluble. Other solubilities: solubel in ethanol, chloroform, acetone, glacial, acetic acid
Formula Weight 413.42
Chemical Name or Material (S,R)-Noscapine
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Harmful if swallowed.

Precautionary Statement GHS P Statement IF SWALLOWED: rinse mouth.

Do NOT induce vomiting.

Call a POISON CENTER or doctor/physician if you feel unwell.

Wash face, hands and any exposed skin thoroughly after handling.

EINECSNumber 204-899-2

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.