Learn More
Medchemexpress LLC HY-103598 100mg Medchemexpress, (S,R,S)-AHPC-PEG3-N3 CAS:1797406-80-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY103598100MG
Encompass_Preferred
Medchemexpress, HY-103598 100mg (S,R,S)-AHPC-PEG3-N3 CAS:1797406-80-4 (S,R,S)-AHPC-PEG3-N3 is a synthesized E3 ligase ligand-linker conjugate that incorporates the (S,R,S)-AHPC based VHL ligand and 3-unit PEG linker used in PROTAC technology. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8133
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.