Learn More
Chemglass Life Sciences Stainless Steel Beaker Tong, 15" OAL
Supplier: Chemglass Life Sciences CG1105100
Stainless Steel Beaker Tong, 15" OAL. Tongs have an overall length of 15" with the jaws being covered with ceramic fabric. Configured with a convenient thumb grip to ensure for a firm hold. Minimum grip diameter is ~1-3/4¿.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.