Learn More
AdipoGen (S,S)-Ethylenediamine-N,N'-disuccinic acid trisodium salt solution, 178949-82-1, MFCD20529176, 100 mL
SDP
Supplier:  AdipoGen CDXE0016L100
Encompass_Preferred
MF: C10H13N2Na3O8, MW: 358.19, ~35% in H2O, Appearance: Pale, clear liquid. Aminopolycarboxylic acid. A powerful, readily biodegradable chelant. Complexing agent, exhibiting superior biological degradation (environmentally friendly) compared to EDTA. Used in cosmetic formulations, as effective anti-oxidant and water hardness control, to reduce radical generation, as effective peroxide stabiliser, as preservative potentiator improving piocide efficacy and excellent transition metal chelator.
Catalog No. 50-114-7085
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.