Learn More
Perkin Elmer US LLC Stainless Steel High Pressure Capsules - 30 µL, Qty. 5
Supplier:  Perkin Elmer US LLC B0182901
Encompass_Preferred
High pressure capsules are recommended whenever higher pressures are expected. The covers of these capsules contain a bursting disk that allow sample to escape in the event that pressure exceeds 150 atm. This allows the endothermic signal produced by volatile samples or by-products to be suppressed, which permits the study of explosive materials and can be used in any situation where the a self-generating atmosphere is used. Capsules are available in stainless steel, gold-plated stainles steel, and titanium.
Catalog No. 50-904-2294
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.