Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S,S)-(-)-Hydrobenzoin 99.0+%, TCI America™
SDP

Catalog No. H082425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H082425G 25 g
H08245G 5 g
H0824100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. H082425G Supplier TCI America Supplier No. H082425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2325-10-2
Molecular Formula C14H14O2
Molecular Weight (g/mol) 214.26
MDL Number MFCD00064255
InChI Key IHPDTPWNFBQHEB-KBPBESRZSA-N
Synonym s,s---hydrobenzoin, 1s,2s-1,2-diphenylethane-1,2-diol, s,s-hydrobenzoin, s,s---1,2-diphenyl-1,2-ethanediol, hydrobenzoin,-, unii-wx45q7714b, 1,2-ethanediol, 1,2-diphenyl-, 1s,2s, s,s-1,2-diphenylethylene glycol, 1s,2s---1,2-diphenyl-1,2-ethanediol, isohydrobenzoin
PubChem CID 853020
IUPAC Name (1S,2S)-1,2-diphenylethane-1,2-diol
SMILES C1=CC=C(C=C1)C(C(C2=CC=CC=C2)O)O

Specifications

Melting Point 148°C
Color White
Quantity 25 g
Formula Weight 214.26
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (S,S)-(-)-Hydrobenzoin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.