Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ S-Sulfo-L-cysteine sodium salt
GREENER_CHOICE

Catalog No. 016210
Change view
Click to view available options
Quantity:
10 mg
50 mg
Catalog No. Quantity
01-621-0 10 mg
01-625-0 50 mg
2 options
This item is not returnable. View return policy

Catalog No. 01-621-0

Supplier: Tocris Bioscience™ 0162/10

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Group I agonist

S-Sulfo-L-cysteine sodium salt is an EAA receptor agonist. Agonist at mGlu1α and mGlu5a subtypes expressed in clonal RGT cell lines.

Specifications

CAS 7381-67-1
Quantity 10 mg
Solubility Soluble to 100 mM in water
Molecular Formula C3H6NO5S2Na
Formula Weight Observed MW: 223.19

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.