Learn More
Abcam S-Sulfocysteine Sodium Salt, Glutamate Receptor Agonist, Solid, 10 mg
Description
- Molecular Weight: 223.2 Da
- Form: Solid
- Activity: Glutamate receptor agonist and glutamate analog
- Biological activity: Precursor to L-cysteine; assumed to cause NMDA receptor-mediated neurodegeneration in molybdenum cofactor deficiency (MoCD) patients; may be involved in cysteine-induced neurotoxicity
- Storage: Store at -20°C
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Content And Storage | Store at -20°C. |
| Shipping Condition | Ambient |
| Substrate Properties | Chemical Substrate |
| Form | Solid |
| Target Molecule | Glutamate Receptor |
| Quantity | 10 mg |
| For Use With (Application) | Glutamate receptor agonist |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.