Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abcam S-Sulfocysteine Sodium Salt, Glutamate Receptor Agonist, Solid, 50 mg
Description
- Molecular Weight: 223.2 Da
- Form: Solid
- Activity: Glutamate receptor agonist and glutamate analog
- Biological Function: Precursor to L-cysteine; involved in cysteine-induced neurotoxicity and NMDA receptor-mediated neurodegeneration in molybdenum cofactor deficiency (MoCD) patients
- Composition: 100% S-Sulfocysteine sodium salt
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | Glutamate Receptor |
| Shipping Condition | Ambient |
| Substrate Properties | Chemical Substrate |
| Form | Solid |
| Buffer | 100% S-Sulfocysteine sodium salt |
| Content And Storage | Store at -20°C. |
| Formulation | 100% S-Sulfocysteine sodium salt |
| Quantity | 50 mg |
| For Use With (Application) | Glutamate receptor agonist |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?