Learn More
Enzo Life Sciences S-Trityl-L-cysteine (500mg). CAS: 2799-07-7
Supplier: Enzo Life Sciences ALX105011M500
Potent, cell permeable cysteine thioether displaying anti-mitotic and anti-tumor properties. Inhibits mitosis by blocking basal and microtubule-activated ATPase activities of the mitotic kinesin Eg5 (IC50=1µM and 140nm). Induces mitotic arrest in HeLa cells (IC50=700nM) and inhibits the growth of NCI tumor cell line panel (GI50=1.31µM). Alternative name: NSC 83265, STLC. Formula: C22H21NO2S. MW: 363.5. Purity: ≥98% (TLC). Appearance: White to off-white solid. Solubility: Soluble in DMSO or acetic acid. Long Term Storage: -20°C.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.